missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Monomeric Avidin Agarose

Catalog No. PI20228
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20228 5 mL
PI20267 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20228 Supplier Thermo Scientific™ Supplier No. LSG20228

Reversibly bind biotinylated proteins and other ligands with monomers of avidin covalently attached as individual subunits onto beaded resins.

Thermo Scientific™ Pierce Monomeric Avidin Resins and Kit enable gentle, reversible-binding and purification of biotinylated proteins.

Pierce Monomeric Avidin Kit and Resins are unique among biotin-binding supports in allowing non-denaturing affinity purification of biotinylated molecules. The immobilized monomeric avidin protein binds biotin with high specificity and moderate affinity, allowing non-biotin molecules to be washed away and then the bound biotin-labeled molecules to be competitively eluted using 2mM biotin in phosphate buffered saline (PBS). This technique provides the gentlest elution conditions for biotinylated protein purification and avoids the contamination and other problems associated with traditional avidin and streptavidin methods. The monomeric avidin column can then be regenerated by using 0.1M glycine to strip the column of residual bound biotin without losing the ability to bind another biotinylated sample.

With typical avidin or streptavidin, the biotin-binding affinity (Kd = 10-15M) is so great that purification with these traditional media require denaturing conditions for elution, such as 8M Guanidine·HCl at pH 1.5 or boiling in reducing SDS-PAGE sample loading buffer. In addition to their adverse effects on the biotinylated protein of interest, such harsh elution conditions typically cause extraneous proteins (i.e., the avidin or streptavidin subunits) to strip from the resin and co-elute with the desired purification product. By contrast, when avidin is coupled to a resin (e.g., crosslinked beaded agarose or UltraLink Resin) as the subunit monomer, its specificity for biotin is retained, but its biotin-binding affinity decreases to a level (Kd = 10-8M ) that is conducive to subsequent elution.

Highlights:

  • Non-denaturing – purifies biotinylated products using mild elution conditions (2mM free biotin)

  • Reusable – Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)

  • Specific – retains biotin-binding specificity and low nonspecific binding of native avidin

  • Flexible – bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2mM biotin

  • Good binding capacity – greater than 1.2mg biotinylated BSA/mL of resin

  • Convenient – purchase standalone agarose or UltraLink™ Resins or a complete purification kit with prepacked column and buffers

Item-Specific Features:

  • Agarose Resins and Kit are based on crosslinked 4% beaded agarose
  • UltraLink Resin is an acrylamide support (UltraLink Biosupport)

Includes:

Kit contains a pre-packed column of Monomeric Avidin Agarose and binding and elution buffers.

Specifications

Capacity (Metric) ≥1.2 mg Biotinylated BSA Binding per mL Resin
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Product Line Pierce
Quantity 5 mL
Target Biotin-Tagged Molecule
Type Agarose
Stationary Phase Avidin
Format Bottle
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.