missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Spin Columns - Screw Cap

Catalog No. PI69705
Change view
Click to view available options
Quantity:
25 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI69705 25 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI69705 Supplier Thermo Scientific™ Supplier No. LSG69705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform small-scale affinity methods, such as immunoprecipitation assays with agarose beads using these empty spin columns and cups (5 to 500μL).

Thermo Scientific™ Pierce Microcentrifuge Spin Cups and Spin Columns are convenient tools for manipulating small volumes of affinity supports for protein purification.

Additional Information and Online Ordering

These microcentrifuge spin cups and spin columns are convenient tools for manipulating small volumes of affinity supports (5 to 500μL) for immunoprecipitation (IP) and pull-down purification assays. Five styles of polypropylene columns and matching collection tubes offer a variety of options to suit particular types of experimental needs. Add the affinity resin and sample to one of the columns and use a microcentrifuge to efficiently wash away contaminants and elute your purified sample without losing any resin in the process. Spin columns allow you to affinity-purify more protein in less time.

Highlights:

  • Efficient washing of samples means fewer washes are needed to remove contaminating proteins
  • Efficient elution of samples means better recovery of immunoprecipitation and assay products
  • Filter cups and columns eliminate contamination of eluted samples with affinity resin
  • Filter cups and columns eliminate the need to decant supernatant from pelleted debris or resin
  • Spin protocols drastically reduce the time required for small-scale affinity protocols
  • Low protein-binding polypropylene column construction minimizes nonspecific binding

Spin Cups-Paper Filter with Collection Tubes (Part No. 69700):

  • Column volume: 600μL
  • Resin volume: 20 to 300μL
  • Filter type: paper, 10μm pore size
  • Cap type: collection tube cap fits onto inserted spin cup

Spin Cups-Cellulose Acetate Filter with Collection Tubes (Part No. 69702):

  • Column volume: 800μL
  • Resin volume: 20 to 400μL
  • Filter type: cellulose acetate, 0.45μm pore size
  • Cap type: collection tube cap fits onto inserted spin cup
  • Spin ColumnsScrew Cap with Luer-Lok

Spin Columns-Screw Cap (Part No. 69705):

  • Column Volume: 900μL
  • Resin volume: 20 to 400μL
  • Filter type: polyethylene filter, 10μm pore size; small and large frit options for different sample sizes
  • Cap type: O-ring screw top caps; press-in bottom plugs
  • Luer-Lok Adapters enable use with syringe-based kits

Spin Columns-Snap Cap with Collection Tubes (Part No. 69725):

  • Column volume: 1000μL
  • Resin volume: 20 to 500μL
  • Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
  • Cap type: snap cap on column (no cap on collection tube); press-on bottom caps

Micro-Spin Columns (Part No. 89879):

  • Column volume: 400μL
  • Resin volume: 5 to 100μL
  • Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
  • Cap type: O-ring screw top caps; press-on bottom caps

Microcentrifuge Tubes (Part No. 69715):

  • 2mL capacity
  • Dolphin-style bottom with attached snap cap
  • Included and compatible with Part No. 69700

Microcentrifuge Tubes (Part No. 69720):

  • 2mL capacity
  • Short, conical bottom with attached snap cap
  • Included and compatible with Part No. 69702

Specifications

Description Pierce Spin Columns - Screw Cap
Content And Storage Store product at room temperature.
Product Line Pierce
Quantity 25 Columns
Type Spin Column
Format Microcentrifuge, Spin Column
Column Type Empty Column, Polyethylene Filter

For Research Use Only. Not for use in diagnostic procedures.