missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MA(PEG)12 Methyl-PEG-Amine Compound

Catalog No. PI26114
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26114 100 mg
PI26115 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI26114 Supplier Thermo Scientific™ Supplier No. 26114

MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.

Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.

MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.

Highlights:

  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures

Recommended for:

  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis

Specifications

Chemical Reactivity Aldehyde
Content And Storage Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Amine, Primary Amine
Spacer 12 ethylene glycol units
Form Solid
Quantity 100 mg
Solubility Water
Product Type Amine Compound

For Research Use Only. Not for use in diagnostic procedures.