missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MBS (m-maleimidobenzoyl-N-hydroxysuccinimide ester)

Catalog No. pi22311
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22311 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22311 Supplier Thermo Scientific™ Supplier No. LSG22311

Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and maleimide groups at ends of an aromatic, 6-atom (7.3A) spacer arm.

Thermo Scientific™ Pierce MBS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 Ås).

Highlights:

  • Popular for forming enzyme immunoconjugates
  • Reactive groups: NHS-ester and maleimide
  • Reactive toward: amino and sulfhydryl groups

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description MBS
Molecular Weight (g/mol) 314.25
PEGylated No
Spacer Arm Length 7.3 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for MBS (m-maleimidobenzoyl-N-hydroxysuccinimide ester)?

MBS (m-maleimidobenzoyl-N-hydroxysuccinimide ester) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.