missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ beta-Galactosidase Assay Reagent

Catalog No. PI75705
Change view
Click to view available options
Quantity:
25 mL
3 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI75705 25 mL
PI75710 3 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI75705 Supplier Thermo Scientific™ Supplier No. 75705
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lyse cultured mammalian cells and measure beta-galactosidase activity using compatible M-PER Cell Lysis Reagent and colorimetric b-Gal Assay Reagent.

The Thermo Scientific Mammalian β-Galactosidase Assay Reagent is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Features of the Thermo Scientific Mammalian β-Galactosidase Assay Kit (assay reagent and stop solution):

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatible with prepared lysates from other species such as bacteria

Related Products

Mammalian beta-Galactosidase Assay Kit (Cat. No. 75707)

beta-Galactosidase Assay Stop Solution (Cat. No. 75706)

Specifications

Assay Reporter Enzyme
Content And Storage Store at -20°C.
Detection Method Absorbance
Form Liquid
For Use With (Application) Absorbance
Product Line Pierce
Quantity 25 mL
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Format 96-well Plate
For Use With (Equipment) Microplate Reader
Label or Dye Enzyme Labeled
Product Type Pierce beta-Galactosidase Assay Reagent
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.