missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KMUH (N-κ-maleimidoundecanoic acid hydrazide)

Catalog No. pi22111
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22111 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22111 Supplier Thermo Scientific™ Supplier No. LSG22111

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (19.0A) crosslinker containing maleimide and hydrazide groups.

Thermo Scientific™ Pierce KMUH is a long, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Highlights:

  • Long, noncleavable, aliphatic spacer
  • Hydrazide group covalently couples to oxidized carbohydrate residues in glycoproteins and other glycoconjugates
  • Reactive groups: maleimide and hydrazide
  • Reactive toward: sulfhydryl and carbonyl (aldehyde) groups (or carboxyl groups when used with EDC)

Specifications

Chemical Reactivity Carbohydrate-Sulfhydryl
Cleavable No
Description KMUH
Molecular Weight (g/mol) 409.4
PEGylated No
Spacer Arm Length 19.0 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Hydrazide, Maleimide
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for KMUH (N-κ-maleimidoundecanoic acid hydrazide)?

KMUH (N-κ-maleimidoundecanoic acid hydrazide) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.