missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Iodoacetic Acid

Catalog No. PI35603
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI35603 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI35603 Supplier Thermo Scientific™ Supplier No. 35603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.

Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.

Iodoacetic Acid (IAA) can react with several protein functional groups but is typically used for specific S-carboxymethylation of sulfhydryls (reduced cysteines).

Iodoacetic acid reacts with sulfhydryls on cysteines, imidazolyl side chain nitrogens of histidines, the thioether of methionine and the primary amine group of lysines.The rate of reaction and specificity is dependent on the ionization level, which can be manipulated by the pH of the reaction condition.

Highlights:
  • React at slightly alkaline pH for specific S-carboxymethylation of free sulfhydryls
  • React at low pH for specific carboxymethylation of methionines
  • React at high pH to favor carboxymethylation of histidines and lysines
  • Methylate reduced cysteine peptide fragments in protease digests for mass spectrometry

Specifications

Chemical Reactivity Thiol
Description Iodoacetic Acid
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Label or Dye Unreactive Group
Label Type Blocking Groups
Product Line Pierce
Spacer Short (<10 Å)
Form Powder
Quantity 500 mg
Solubility Water
Product Type Iodoacetic Acid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.