missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Intact Protein Standard Mix

Catalog No. p-7083442
Encompass
Change view
Click to view available options
Quantity:
1 x 76 μg
5 x 76 μg
Catalog No. Quantity
PIA33526 1 x 76 μg
PIA33527 5 x 76 μg
2 options

Catalog No. PIA33526

Supplier: Thermo Scientific™ A33526

Only null left

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Intact Protein Standard Mix
Product Line Pierce
Content And Storage 1 vial, store at -5 to -30°C
Shipping Condition Wet Ice
Final Product Type Intact Proteins
Workflow Step Calibration
Quantity 1 x 76 μg
Form Powder

For research use only. Not for use in diagnostic procedures.