missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Intact Protein Standard Mix

Catalog No. PIA33526
Encompass
Change view
Click to view available options
Quantity:
1 x 76 μg
5 x 76 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA33526 1 x 76 μg
PIA33527 5 x 76 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA33526 Supplier Thermo Scientific™ Supplier No. A33526

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Intact Protein Standard Mix
Product Line Pierce
Content And Storage 1 vial, store at -5 to -30°C
Shipping Condition Wet Ice
Final Product Type Intact Proteins
Workflow Step Calibration
Quantity 1 x 76 μg
Form Powder
Which standards and calibrants do you recommend using for assessing LC-MS system suitability?

Here are our recommendations:

- Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321) for LC system and gradient optimization
- Pierce BSA Protein Digest, MS grade (Cat. No. 88341), Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342), and Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) for "bottom up" LC-MS method optimization and LC-MS suitability assessment
- Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) (Cat. No. A40010) for assessing the gradient, sensitivity, and dynamic range of the LC-MS system for discovery and targeted peptide quantitative workflows
- Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for assessing system performance for TMT-based quantitative workflows
- Pierce Intact Protein Standard Mix (Cat. Nos. A33526, A33527) for "top down" LC-MS method optimization and LC-MS suitability assessment

For research use only. Not for use in diagnostic procedures.