missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immobilized Papain (Agarose Resin)

Catalog No. PI20341
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20341 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20341 Supplier Thermo Scientific™ Supplier No. 20341

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Generate and purify Fab fragments from antibodies with cysteine-endopeptidase papain immobilized onto beaded agarose resin.

Thermo Scientific™ Pierce Immobilized Papain consists of a cysteine-endopeptidase (papain) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab fragments from antibodies.

Papain is a nonspecific, cysteine-endopeptidase isolated from papaya latex that is used in a wide variety of applications. When IgG molecules are incubated with papain in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing three fragments of similar size: two Fab fragments and one Fc fragment. The Fc fragments can be separated from the Fab fragments using immobilized Protein A or by ion exchange chromatography. The 50 kilodalton Fc fragment is often used for determining specificity of the Fc receptors without interference from antigen binding. Fab fragments are useful in immunohistochemical studies because the fragments penetrate the tissue better than intact IgG. Also, nonspecific binding from Fc receptors is avoided by using only the Fab portion of the immunoglobulin. Papain can als be used to generate F(ab')2 fragments. To prepare F(ab')2 fragments, the papain is first activated with 10mM cysteine, which is then removed via gel filtration. If no cysteine is present during the papain digestion, F(ab')2 fragments can be generated. These fragments are often inconsistent and reproducibility is a problem. Immobilized papain can be substituted for free papain in almost any application, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the papain or controlling the flow of sample over an immobilized papain column. Immobilized papain is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.

Highlights:

  • Support: Crosslinked 6% beaded agarose
  • Specific Activity: 16-40 BAEE units/mL papain before loading
  • Loading: 250μg papain/mL gel
  • Form: Slurry in 50% glycerol, 0.1M Na-acetate, pH 4.4, 0.05% sodium azide

Additional Features:

  • Alternate name: Papaya peptidase I
  • Cleaves somewhat nonspecifically at exposed residues
  • Source: Carica papaya latex Molecular weight: 23,000
  • Protease type: Cysteine protease
  • For antibody fragmentation, cleaves into Fab or F(ab')2 fragments
  • Reaction conditions pH 6 to 7, 20 to 37°C
  • Inhibitors: heavy metals, carbonyls, NEM, p-chloromercuro-benzoate, E-64

Specifications

Content And Storage Upon receipt store at 4°C.
Form Liquid Suspension
Quantity 5 mL
Product Type Papain
Can I purchase the immobilized enzymes (proteases) from the Thermo Scientific IgG Fragmentation kit separately?

Yes. The three immobilized enzymes are available in 5 mL package sizes as shown below:

• Immobilized Ficin, Cat. No. 44881
• Immobilized Papain, Cat. No. 20341
• Immobilized Pepsin, Cat. No. 20343

How much of the immobilized Papain (Agarose Resin) do I need for digestion of 10 mg IgG?

0.5 mL of the 50% Immobilized Papain slurry is sufficient for the digestion of up to 10 mg of pure lyophilized IgG reconstituted in 1 mL of Digestion Buffer.

For Research Use Only. Not for use in diagnostic procedures.