missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Hydroxylamine-HCl

Catalog No. PI26103
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26103 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26103 Supplier Thermo Scientific™ Supplier No. 26103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NH2OH (CAS 7803-49-8) to cleave Asn-Gly peptide bonds or deprotect sulfhydryls following S-acetylthioacetate modification with SATA reagent.

Thermo Scientific™ Pierce Hydroxylamine-HCl has many uses, including to cleave Asn-Gly peptide bonds and to deprotect (expose) sulfhydryls for conjugation following modification with SATA and similar reagents.

Thermo Scientific™ Pierce Hydroxylamine-HCl has many uses, which include cleaving Asn-Gly peptide bonds and deprotecting (exposing) sulfhydryls on SATA-modified molecules for conjugation. Hydroxylamine·HCl is a strong reducing agent that is useful in biochemical crosslinking applications, including the deacetylation of SATA and chemical cleavage of EGS and Sulfo-EGS. Hydroxylamine converts carbonyl compounds (aldehydes and ketones) to their oxime derivatives in the presence of a weak base. Therefore, crosslinkers and other compounds that contain a carbonyl within their structure are cleavable with hydroxylamine-HCl.

Highlights:

  • Exposes protected sulfhydryl groups on SATA-modified molecules
  • Cleaves carbonyl-containing crosslinkers such as EGS

Reagent Properties:

  • Chemical name: Hydroxylamine Hydrochloride
  • Chemical formula: NH2OH·HCl
  • Molecular weight: 69.49
  • CAS Number: 5470-11-1
  • Form: White crystals, free of foreign material
  • Solubility: 10%

Specifications

CAS 5470-11-1
Content And Storage Upon receipt store desiccated at room temperature. Product shipped at ambient temperature.
Product Line Pierce
Form Solid
Reagent Type SATA
Quantity 25 g
Product Type Hydroxylamine Hydrochloride

For Research Use Only. Not for use in diagnostic procedures.