missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL)

Catalog No. PI88947
Change view
Click to view available options
Quantity:
10,000 U
1000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88947 10,000 U
PI88946 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88947 Supplier Thermo Scientific™ Supplier No. 88947

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Remove fusion tags from proteins with the HRV 3C cleavage sequence with this recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.

Remove fusion tags from proteins with the HRV 3C cleavage sequence with Thermo Scientific™ Pierce Human Rhinovirus (HRV) 3C Protease, a recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.

Additional Information and Online Ordering

Pierce HRV 3C Protease is highly specific for the recognition sequence Leu-Glu-Val-Leu-Phe-Gln▾Gly-Pro and cleaves after the glutamine residue. The recombinant enzyme is engineered as a fusion protein with glutathione-S-transferase (GST) and polyhistidine (6xHis) tags, so that the protease can be easily removed after cleavage using a variety of solid supports with immobilized glutathione (GSH) or metal chelates, respectively. HRV3C is also active in a variety of commonly used buffers, providing further flexibility in experimental design.

Highlights:

  • Effective – HRV 3C is designed to remove purification tags from your protein of interest

  • High activity at 4°

    C – activity is ≥2000U/mg control protein; one unit will cleave 0.1mg control protein at 4°C after 16h

  • Flexible – dual-tagged enzyme is compatible with various columns and buffers

Includes:

Kit contains protease, cleavage buffer and control protein.

Recommended for:

Removal of purification tags from fusion proteins

Specifications

Content And Storage Store at -20°C.
Product Line Pierce
Form Kit
Quantity 10,000 U
Product Type Protease

For Research Use Only. Not for use in diagnostic procedures.