missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity NeutrAvidin™ Agarose

Catalog No. p-7184469
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI29202 5 mL
PI29204 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI29202 Supplier Thermo Scientific™ Supplier No. 29202

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylamide and agarose beads and columns of specially-modified, pH-stabilized avidin; the low-cost alternative to streptavidin biotin-binding supports.

Thermo Scientific™ Pierce NeutrAvidin Resins provide less nonspecific binding and cleaner results than avidin or streptavidin for many kinds of biotin affinity procedures.

Additional Information and Online Ordering

NeutrAvidin Agarose and UltraLink™ Resins are superior alternatives to avidin and streptavidin beads in many biotin-based affinity applications. NeutrAvidin Protein is a modified avidin derivative that combines several key features to provide a biotin-binding protein with exceptionally low nonspecific binding properties. Native avidin is heavily glycosylated, resulting in its high pI and nonspecific binding properties. By contrast NeutrAvidin Protein contains no carbohydrate and has a near-neutral isoelectric point (6.3), features that provide for very low nonspecific binding and broader utility in a variety of applications.

NeutrAvidin Resins are prepared by covalently coupling NeutrAvidin Protein to the beaded agarose or UltraLink Biosupport using efficient and stable chemistries, resulting in NeutrAvidin Agarose and UltraLink Resins that are leach-resistant and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Highlights:

  • Low nonspecific binding – unlike avidin, NeutrAvidin Protein has no carbohydrates, eliminating nonspecific binding problems caused by sugars; unlike streptavidin, NeutrAvidin Protein has no Arg-Tyr-Asp sequence, which can be a source of binding to cell surface molecules

  • Versatile resins – agarose (polysaccharide-based) and UltraLink Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods

  • Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)

  • Superior performance – High Capacity Resin has higher binding capacity than best alternative supplier streptavidin products, allowing use of small amounts of resin for experiments

Item-Specific Features:

  • Agarose Resins are crosslinked 6% beaded agarose
  • UltraLink Resins are beaded acrylamide support (UltraLink Biosupport)
  • Plus and High Capacity Resins have a higher loading of NeutrAvidin Protein than the regular forms

Recommended for:

Immunoprecipitation; Purifying proteins that bind to biotinylated ligands; Capturing biotinylated cell-surface proteins; Purifying biotinylated peptides

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature.
Particle Size 45 to 165 μm
Product Line NeutrAvidin, Pierce
Quantity 5 mL
Target Biotin-Tagged Molecule
Type Agarose
Stationary Phase NeutrAvidin Protein
Format Bottle
Column Type Affinity, Agarose Resin
Binding Capacity 8 mg Biotinylated BSA Binding/mL Resin

For Research Use Only. Not for use in diagnostic procedures.