Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Description
Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.
Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Specifications
| Capacity (Metric) | Up to 2,000,000 EU/mL |
| Formulation | 22mL-capacity centrifuge columns containing 1mL resin |
| Content And Storage | Upon receipt store at 4°C. |
| Form | Liquid |
| Product Line | Pierce |
| Quantity | 5 Columns |
| Target | Endotoxin Removal |
| Type | Spin Column |
| Stationary Phase | Proprietary Ligand |
| Format | Spin Column |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.