Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Description
Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour; with this spin cup format, protein recovery is ≥85%.
Features of High Capacity Endotoxin Removal Resin Spin Columns:
- High capacity—bind up to 2,000,000 EU/mL† to eliminate >99% of endotoxins
- Selective—recover ≥85% of your protein sample
- High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
- Fast—our new spin cup format enables endotoxin depletion within 1 hour
- Clean—single-use spin columns avoid cross contamination of samples
- Optimized—spin cups are optimized for different sample volumes
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.
Requires:
- Endotoxin-free water
Applications:
- Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.
Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for >85% protein recovery.
† One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.
Related Products
- Pierce™ High Capacity Endotoxin Removal Resin (Cat. No. 88270)
- Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL (Cat. No. 88273)
- Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL (Cat. No. 88274)
Specifications
Specifications
| Formulation | 22 mL-capacity centrifuge columns containing 1 mL resin |
| Content And Storage | Upon receipt store at 4°C. |
| Form | Liquid |
| Product Line | Pierce |
| Quantity | 5 Columns |
| Target | Endotoxin Removal |
| Type | Spin Column |
| Stationary Phase | Proprietary Ligand |
| Format | Spin Column |
| Column Type | Affinity, Cellulose Resin |
| Show More |
Frequently Asked Questions (FAQs)
We have not tested this, but the resin is autoclavable up to 121 degrees C at 1 bar for 20 min.
For Research Use Only. Not for use in diagnostic procedures.