missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Resin

Catalog No. pi88271
Change view
Click to view available options
Quantity:
10 mL
100 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88271 100 mL
PI88270 10 mL
PI88272 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI88271 Supplier Thermo Scientific™ Supplier No. 88271
Only null left

Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.

Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour while maximizing protein recovery. Use this endotoxin removal resin slurry to pack a custom column that can be re-used up to 10 times with no loss in endotoxin removal efficiency.

Features of High Capacity Endotoxin Removal Resin:

  • High capacity—bind up to 2,000,000 EU/mL to eliminate >99% of endotoxins
  • Durable—re-use resin up to 10 times with no loss in performance
  • Selective—recover ≥85% of your protein sample
  • High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
  • Economical—large-volume discounts available

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by ≥99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.

Requires:

Endotoxin-free water

Applications:

Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.

Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This resin is offered in a slurry format for custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10-15 mL/hour.

One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.

Related Products

  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL (Cat. No. 88273)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL (Cat. No. 88274)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL (Cat. No. 88276)

Specifications

Capacity (Metric) Up to 2,000,000 EU/mL
Formulation Proprietary ligand on beaded cellulose slurried in 20% ethanol
Content And Storage Upon receipt store at 4°C.
Form Liquid
Product Line Pierce
Quantity 100 mL
Target Endotoxin Removal
Type Resin
Stationary Phase Proprietary Ligand
Format Bottle
Column Type Affinity, Proprietary Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.