missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Guanidine-HCl

Catalog No. PI24110
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24110 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI24110 Supplier Thermo Scientific™ Supplier No. 24110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pure, crystalline guanidium hydrochloride (GdnHCl, GuHCl), CAS 50-01-1; denaturant for protein biology methods (elution, solubilization, refolding, storage).

Thermo Scientific™ Pierce Guanidine-HCl is pure, crystalline guanidine hydrochloride, a denaturant with many protein research applications.

Additional Information and Online Ordering

Highlights:

  • Guanidine·HCl (Sequanal grade) is a highly purified, ready-to-use denaturant
  • Free of UV-absorbing materials in the range of 225 – 300nm
  • Sharp UV cut-off spectrum with A260 less than 0.03

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Form Powder
Reagent Type Chaotrope, Denaturant
Quantity 500 g
Product Type Guanidine Hydrochloride

For Research Use Only. Not for use in diagnostic procedures.