missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit

Easily capture and affinity purify binding partners of GST-tagged fusion proteins with these glutathione (GSH) agarose beads and complete reagent set.

Supplier:  Thermo Scientific™ LSG21516

Catalog No. PI21516


Only null left
Explore available promotions

Description

Description

Thermo Fisher Scientific Pierce GST Tag Protein Interaction Pull-Down Kits contains the necessary components to capture and purify proteins that interact with GST-tagged fusion proteins.

Features of GST Tag Protein Interaction Pull-Down Kit:

  • GST pull-down (Product No. 21516)—gently purifies protein interactors of any GST-tagged fusion protein without denaturing
  • Complete kits—provide all components and detailed protocol for purifying protein:protein interactions
  • No special equipment needed—use common laboratory equipment and reagents (e.g., microcentrifuge)
  • Convenient—microcentrifuge spin columns facilitate simple and efficient manipulation of agarose beads, including simple processing of multiple samples
  • Flexible—instructions include protocols for use with bait and prey proteins from many different sources
  • Binding—Binds approx. 8 mg of GST-tagged fusion protein per mL of resin

Applications:

  • Discover a new protein:protein interaction from a cell lysate
  • Confirm a putative interaction from a cell lysate or with a previously purified protein
  • Extract protein:protein interaction information from in vitro transcription/translation lysates

You provide the tagged fusion protein as the 'bait' and the cells expressing the putative protein interaction target ('prey'), and the Pull-Down Kits provide everything else: cell lysis buffer, microcentrifuge spin columns, tag-specific affinity resin (agarose beads) and optimized buffers and protocol. The Pull-Down Kits are designed to teach the method to first-time users and to increase ease-of-use, convenience and reproducibility for experienced researchers.

Related Products

Pierce™ His Protein Interaction Pull-Down Kit (Catalog #21277)

Specifications

Specifications

750 μL
Slurry: 50 %
Affinity Purification
Pierce
Pull-Down (Tag-Based)
Pull-Down Kit
Kit
1.5 mL
Sufficient For: At least 25 pull-down reactions, each using 25 μL of resin
• Glutathione Agarose, 750 μL
• Pull-Down Lysis Buffer, 250 mL
• Glutathione, 1 g
• TBS Pack (makes 500 mL), 1 pack
• Pierce Spin Columns and Accessories, 27 columns
• Microcentrifuge Collection Tubes, 100 tubes

Upon receipt store Pierce Lysis Buffer at room temperature. Store remaining kit components at 4°C.
Microcentrifuge
25 reactions kit
Not Target-Specific
25 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.