Learn More
Thermo Scientific™ Pierce™ GST Spin Purification Kit, 1 mL
Description
Thermo Scientific™ Pierce Glutathione Agarose is a high-capacity, high-performance resin for affinity purification of GST-tagged fusion proteins from cellular lysates.
To accommodate these many uses, Pierce Glutathione Agarose is offered in several package sizes and formats. These include three volumes of resin slurry, three sizes of centrifuge-ready columns, complete GST purification kits, and two sizes of FPLC–ready chromatography cartridges.
Highlights:
High capacity – binds at least 25mg of recombinant GST protein per milliliter of resin
High yield and purity – consistently purifies at least 10mg of GST – tagged protein per milliliter of resin with greater than 90% purity
Cost-effective – resin is economically priced and can be reused at least five times without reduction in binding capacity and purification performance
Versatile – works well to purify GST – fusion proteins from bacterial lysates or use with pre-purified GST – tagged proteins to pull down protein interactions
Compatible – validated and effective for use with Thermo Scientific Pierce Cell Lysis Reagents to extract and purify from bacterial or mammalian cell cultures
Flexible – available in multiple formats including bulk resin, spin columns, chromatography cartridges and convenient GST purification kits
Available Formats:
Resin Slurries: 10mL, 100mL, 500mL bottles
Spin Columns: 0.2mL (microcentrifuge), 1mL, 3mL columns
Purification Kits: complete kits in all three column sizes
Chromatography Cartridges: 1mL and 5mL cartridges, compatible with automated liquid chromatography systems and with syringe processing
Bulk and Custom: let us know your needs for bulk Glutathione Agarose
Includes:
Kits contain Pierce Glutathione Spin Columns, equilibration/wash buffer and glutathione to prepare elution buffer
Specifications
Specifications
| Capacity (Metric) | 1 mL |
| Description | Pierce GST Spin Purification Kit, 1mL |
| Content And Storage | Sufficient For: Purifying approx. 10 mg of GST-tagged protein per column • Pierce Glutathione Spin Columns, 1 mL, 5 columns • Equilibration/Wash Buffer, 250 mL • Glutathione, 184 mg • Column Plugs, 5 plugs Store at 4°C. |
| Product Line | Pierce |
| Quantity | 5 Column Kit |
| Target | GST-Tagged Protein |
| Type | Spin Column Kit |
| Stationary Phase | Glutathione |
| Sufficient For | 10 mg of GST-Tagged Protein per Column |
| Format | Spin Column |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.