missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution

Catalog No. PIA39239
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA39239 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA39239 Supplier Thermo Scientific™ Supplier No. A39239

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution is a mixture of 16 highly pure, ionizable components designed for both positive and negative ionization calibration using Orbitrap Tribrid Series 3.1 Instrument Control Software.

  • Improved mass accuracy - extended mass range improves mass accuracy
  • Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
  • Highly validated - high-quality, rigorously-tested formulation with lot-specific certificates of analysis
  • Ready-to-use - reference standard may be directly loaded into a syringe and injected into the instrument
  • Store at room temperature for up to 18 months

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Calibration Solution
Chemical Reactivity Amine
Product Line Pierce
Shipping Condition Room Temperature
Workflow Step Calibration
Quantity 10 mL
Form Liquid
What is the difference between Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?

Both the Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers have the same 16 components and produce the same spectra. However, the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers has a different storage solution: LC-MS grade acetonitrilie 87% (v/v) and LC-MS grade water 13% (v/v). The FlexMix Calibration for Auto-Ready Mass Spectrometers is designed to be used only in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).

What type of instruments does the Pierce FlexMix Calibration Solution work with?

We recommend using the Pierce FlexMix Calibration Solution with Orbitrap Fusion, Orbitrap Fusion Lumos, and Orbitrap ID-X MS instruments.

What is the Pierce FlexMix Calibration Solution, and what is its main purpose?

The Pierce FlexMix Calibration Solution is a mixture of 16 highly pure, ionizable components (mass ranges: 50 to 3,000 m/z) designed for both positive and negative ionization calibration using Orbitrap Tribrid Series 3.1 Instrument Control Software (or later) for Orbitrap Fusion, Orbitrap Fusion Lumos, and Orbitrap ID-X MS instruments.

For Research Use Only. Not for use in diagnostic procedures.