missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Blocker Blocking Buffer

Catalog No. ENN502
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENN502 Supplier Thermo Scientific™ Supplier No. N502
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increases signal-to-noise ratio and minimizes background

Reagent sold ready-to-use.

Specifications

Product Type Blocking Buffer
Quantity 1 L
Form Liquid
For Use With (Application) ELISA
Content And Storage Store at 2–8°C.

For Research Use Only. Not for use in diagnostic procedures.