missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EDTA

Catalog No. PI17892
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17892 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17892 Supplier Thermo Scientific™ Supplier No. LSG17892

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chelate alkaline earth metals. Thermo Scientific™ Pierce EDTA is highly purified for use in a variety of laboratory methods.

Thermo Scientific™ Pierce EDTA is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc in a variety of laboratory methods.

Additional Information and Online Ordering

EDTA (ethylenediaminetetraacetic acid) is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc. EDTA is especially useful for its ability to sequester metal ions to reduce their reactivity in solutions.

  • White crystalline powder
  • ≥99.0% Pure
  • Water soluble
  • MW: 372.24 g/mol

Specifications

Chemical Name or Material EDTA
CAS 6381-92-6
Content And Storage Store at room temperature.
Molecular Formula C10H14N2Na2O8·2H2O
Product Type EDTA (ethylenediaminetetraacetic acid)
Purity ≥99%
Quantity 500 g
Synonym EDTA disodium salt dihydrate
Form Powder
Can I use EDTA (Cat. No. 17892) in molecular biology? If not, can you provide an alternative?

EDTA (Cat. No. 17892) is not intended for molecular biology applications.

UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O) (Cat. No. 15576028) is a potential alternative intended for molecular biology applications.

Do you offer any of the components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets individually?

Yes, the following components of the Pierce protease, phosphatase and (protease and phosphatase) inhibitor tablets and mini tablets are offered separately:

- AEBSF (Cat. No. 78431)
- Aprotinin (Cat. No. 78432)
- Bestatin (Cat. No. 8433)
- E-64 (Cat. No. 78434)
- Leupeptin (Cat. No. 78435)
- Pepstatin A (Cat. No. 78436)
- EDTA (Cat. No. 17892).

For Research Use Only. Not for use in diagnostic procedures.