missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ EDC, No-Weigh™ Format

Catalog No. pia35391
Change view
Click to view available options
Quantity:
10 mg
10 x 1 mg
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA35391 10 x 1 mg
PI77149 10 mg
PI22980 5 g
PI22981 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PIA35391 Supplier Thermo Scientific™ Supplier No. A35391
Only null left

Immobilize or conjugate proteins using 1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide (CAS 25952-53-8), a water-soluble, carboxyl-to-amine crosslinker.

Thermo Scientific™ Pierce EDC is a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation.

Thermo Scientific No-Weigh products are specialty reagents provided in pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive group: Carbodiimide
  • Reaction target: Activates carboxyl groups to conjugate to amino groups (primary amines)
  • Several conjugation strategies: React EDC alone with target groups or include NHS or Sulfo-NHS to increase reaction efficiency or to stabilize active intermediate for later reaction to amines
  • Neutral linkage: Forms neutral amide bonds between carboxyls and amines
  • Water-soluble Reagent: Add directly to reactions in aqueous, physiological buffers
  • Soluble reaction byproducts: Easily removed by washing with water or dilute acid
  • High-purity, crystalline reagent: Use to create high-quality, activated derivatives
Applications:
  • Conjugate carboxyl and amino groups among peptides and proteins
  • Couple haptens to immunogenic carrier proteins (e.g., attach a peptide to KLH)
  • Immobilize peptide antigens to affinity-purify antibodies
  • Create NHS-activated, amine-reactive labeling compounds
  • Crosslink proteins to carboxyl coated beads or surfaces
  • Activate nanoparticles with amine-reactive Sulfo-NHS esters

Specifications

Chemical Reactivity Amine-Carboxyl
Cleavable No
Description EDC
Molecular Weight (g/mol) 191.7
PEGylated No
Spacer Arm Length 0.0 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Wet Ice
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short (<10 Å)
Form Powder
Quantity 10 x 1 mg
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.