missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ EDC, No-Weigh™ Format

Catalog No. pi22980
Change view
Click to view available options
Quantity:
10 mg
10 x 1 mg
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22980 5 g
PIA35391 10 x 1 mg
PI77149 10 mg
PI22981 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI22980 Supplier Thermo Scientific™ Supplier No. 22980
Only null left

Immobilize or conjugate proteins using 1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide (CAS 25952-53-8), a water-soluble, carboxyl-to-amine crosslinker.

Thermo Scientific™ Pierce EDC is a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation.

EDC is the abbreviation for 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, a crosslinker that has been used in diverse applications such as forming amide bonds in peptide synthesis, attaching haptens to carrier proteins to form immunogens, labeling nucleic acids through 5Æphosphate groups and creating amine-reactive NHS-esters of biomolecules. EDC reacts with a carboxyl to form an amine-reactive O-acylisourea intermediate. If this intermediate does not encounter an amine, it will hydrolyze and regenerate the carboxyl group. In the presence of N-hydroxysulfosuccinimide (Sulfo-NHS, Part No. 24510), EDC can be used to convert carboxyl groups to amine-reactive Sulfo-NHS esters.

Highlights:

  • Zero-length crosslinker
  • Numerous chemical conjugates can be synthesized via available –COOH and –NH2 groups
  • Amide bond formed provides a neutral linkage, which is ideal for preparing peptides and antigens
  • Adding Sulfo-NHS (Part No. 24510) enhances the coupling reaction at physiological pH conditions
  • Reactive groups: carbodiimide
  • Reactive toward: carboxyl and amino groups

Specifications

Chemical Reactivity Amine-Carboxyl
Cleavable No
Description EDC
Molecular Weight (g/mol) 191.7
PEGylated No
Spacer Arm Length 0.0 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Wet Ice
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short (<10 Å)
Form Powder
Quantity 5 g
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.