missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ DSP, No-Weigh™ Format

Catalog No. p-4530989
Change view
Click to view available options
Quantity:
50 mg
10 x 1 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA35393 10 x 1 mg
PI22586 50 mg
PI22585 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIA35393 Supplier Thermo Scientific™ Supplier No. A35393
Only null left

Crosslink amines to amines with dithiobis succinimidyl propionate, composed of NHS-ester groups and disulfide-containing, 12.0A spacer arm and cleavable with reducing agents.

Thermo Scientific™ Pierce DSP Crosslinker (Lomant's Reagent) is an amine-reactive crosslinker with NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Amine-reactive NHS ester reacts rapidly with any primary amine-containing molecule
  • Disulfide bond in the spacer arm is readily cleaved by 10-50mM DTT or TCEP at pH 8.5
  • Spacer arm also cleavable with reducing SDS-PAGE sample loading buffer
  • Cleavable crosslinker allows separation of crosslinked products
  • Water-insoluble (dissolve first in DMF or DMSO); compare to DTSSP (Sulfo-DSP)
  • Membrane-permeable, allowing for intracellular crosslinking
Applications:
  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • "Fix" protein interactions to allow identification of weak or transient protein interactions
  • Fix tissues for immunostaining without damaging RNA
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
  • Examining spatial relationships of the capside polypeptides of the mengo virion
  • Studying renal Na+ and K+ -ATPase
  • Nearest neighbor relationships of bovine mitochondrial H+ -ATP
  • Producing interactions between protein components of the chemotaxis mechanism in E. coli
  • Chemical crosslinking of a-CPI

Specifications

Chemical Reactivity Amine-Amine
Cleavable By Thiols
Description DSP
Molecular Weight (g/mol) 404.42
PEGylated No
Spacer Arm Length 12.0 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Wet Ice
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 10 x 1 mg
Solubility DMF, DMSO
Format Standard, Single-use, Premium-grade
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.