missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMS (dimethyl suberimidate)

Catalog No. pi20700
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20700 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20700 Supplier Thermo Scientific™ Supplier No. LSG20700

Crosslink amines to amines with dimethyl suberimidate (CAS 34490-86-3); composed of imidoester groups at either end of a membrane-permeable, 8-atom spacer arm.

Thermo Scientific™ Pierce DMS is dimethyl suberimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of an 8-atom spacer arm.

Highlights:

  • Water-soluble
  • Rapid reaction with amines at alkaline pH values (pH 8-10)
  • Amidine bond retains net charge character of protein
  • Reversible at high pH values
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DMS
Molecular Weight (g/mol) 273.2
PEGylated No
Spacer Arm Length 11.0 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for DMS (dimethyl suberimidate)?

DMS (dimethyl suberimidate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.