missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMP (dimethyl pimelimidate)

Catalog No. p-4531007
Change view
Click to view available options
Quantity:
50 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21666 50 mg
PI21667 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI21666 Supplier Thermo Scientific™ Supplier No. 21666
Only null left

Crosslink amines to amines with dimethyl pimelimidate (CAS 58537-94-3); composed of imidoester groups and a membrane-permeable, 7-atom spacer arm.

Thermo Scientific™ Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.

Highlights:

  • Water-soluble
  • Rapid reaction with amines at alkaline pH values (pH 8-10)
  • Amidine bond retains net charge character of protein
  • Reversible at high pH values
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DMP
Molecular Weight (g/mol) 259.17
PEGylated No
Spacer Arm Length 9.2 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.