missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Disposable Columns, 2 mL

Empty drip columns (0.5 to 5 mL) with matching frits and caps for packing beaded affinity or desalting resins to use in gravity-flow purification techniques.

Supplier:  Thermo Scientific™ 29920

Catalog No. PI29920


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.

Additional Information and Online Ordering

These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.

Highlights:

  • For bench top gravity-flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Columns can be pre-packed and stored until needed
  • Compatible with most types of aqueous buffer eluents commonly used in chromatography
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates

Includes:

Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)

Specifications

Specifications

2 mL
Store at room temperature.
100 columns
Gravity Flow Column
Disposable Columns, 2mL
Pierce
Spin Column
Empty Column, Polyethylene Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.