Learn More
Thermo Scientific™ Pierce™ Cysteine-HCl
Catalog No. PI44889
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
PI44889 5 g
1 options
Catalog No. PI44889 Supplier Thermo Scientific™ Supplier No. 44889
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Make sulfhydryl assay standards or perform protein refolding experiments using this pure, crystalline cysteine hydrochloride (Cys-HCl).
Thermo Scientific™ Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent, or as an additive in protein refolding experiments.
Thermo Scientific™ Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent, or as an additive in protein refolding experiments.
Specifications
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 25°C.
Product Line Pierce
Form Solid
Quantity 5 g
Product Type Cysteine Hydrochloride
For Research Use Only. Not for use in diagnostic procedures.