Learn More
Thermo Scientific™ Pierce™ Controlled Protein-Protein Crosslinking Kit
Description
Thermo Scientific™ Pierce Controlled Protein-Protein Crosslinking Kit contains essential crosslinkers, modification reagents and buffers for conjugating antibodies, enzymes and other purified proteins needed as probes.
Highlights:
Sulfo-SMCC – high-quality crosslinker conjugates primary amines of protein 1 to sulfhydryl groups of protein 2
SATA or 2-MEA – modification reagents provide two options for making sulfhydryl groups available on protein 2 (usually an antibody)
Ellman's Reagent – provides a mean of monitoring and assessing efficiency of the final reaction
Complete – includes critical reagents as well as buffers, desalting columns and supplementary modification reagents
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | Controlled Protein-Protein Crosslinking Kit |
| Molecular Weight (g/mol) | 436.37 |
| PEGylated | No |
| Spacer Arm Length | 11.6 Å |
| Content And Storage | Sufficient For: One conjugation with two purified proteins (up to 5 mg each) • Sulfo-SMCC Crosslinking Agent, 2 mg • Activation/Conjugation Buffer (10X), 20 mL • PBS packs (each makes 500mL), 2 packs • 2-Mercaptoethylamine-HCl, 6 mg • Immobilized-Reductant, 0.2 mL • SATA, 2 mg • Dimethylformamide (DMF), 1 mL • Hydroxylamine-HCl, 5 mg • Dextran Desalting Column, 2 x 10 mL • Column Extender, 2 ea. • Cysteine-HCl, 20 mg • Ellman's Reagent, 2 mg Store at 4°C. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
Frequently Asked Questions (FAQs)
Controlled Protein-Protein Crosslinking Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
For Research Use Only. Not for use in diagnostic procedures.