missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G Coated Plate IP Kit

Catalog No. PI45350
Change view
Click to view available options
Quantity:
2 plate kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI45350 2 plate kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI45350 Supplier Thermo Scientific™ Supplier No. 45350

Perform immunoprecipitation assays without resins, beads, centrifugation or magnets using coated streptavidin or Protein A/G 96-well microplates.

Thermo Scientific™ Pierce Coated Plate IP Kits enable immunoprecipitation experiments to be performed in 96-well microplates.

These coated microplate immuunoprecipiation kits provide for rapid immunoprecipitation of multiple samples without the usual tedium of pipetting, centrifuging and separating beaded affinity resin in individual microcentrifuge tubes. Immunoprecipitation is accomplished using coated 96-well microplates rather than beaded agarose resin. The plate format allows for faster processing of multiple samples. The Protein A/G Coated Plate Kit can be used for immunoprecipation with any antibody that binds to Protein A or G. The Streptavidin Coated Plate Kit is for immunoprecipitation with biotinylated IP antibodies.

Highlights:

  • Ready-to-use, high quality coated plates provide high capacity and consistency
  • Accommodate antibody and sample volumes comparable to tube-based methods
  • Easily recover immunoprecipitated samples for downstream analysis
  • Coated plates are 96-well strip plates, convenient for experiments requiring only a partial plate
  • Include simple instructions, including detailed explanation of appropriate controls
  • Two kits available, suitable for most common antibody types (mouse, rabbit, human and goat IgG subclasses) or any biotinylated antibody or bait protein

Advantages of the Plate Format:

  • Best suited for simultaneously processing multiple samples and their control conditions
  • Allows faster, easier and more thorough washing than with traditional tube/resin IP methods
  • Makes use of familiar and convenient ELISA tools (multichannel pipettors and plate washing)
  • No no tubes to open and close, and no centrifugation required

Protein A/G Kit (Part No. 45350)

  • Binds all human IgG subclasses; human IgM, IgA (weak), and IgE; and all mouse IgG subclasses
  • Offers increased binding for mouse monoclonal antibodies compared to Protein A or Protein G alone
  • Free of pH dependency when binding IgGs, providing more flexible binding conditions

Streptavidin Kit (Part No. 45360)

  • Used in combination with biotinylated antibodies, resulting in IP and co-IP preparations free of interfering antibody fragments
  • Contains biotin-blocking buffer to minimize endogenous biotin binding

Includes:

Coated 96-well strip plates, uncoated 96-well strip plates, PBS pack, detergent solution, elution buffer, neutralization buffer and sealing tape

Recommended for:

Rapid, trouble-free immunoprecipitation of multiple samples and their controls; Immunoprecipitation when plate-based tools and techniques are preferred

Specifications

Capacity (Metric) 2 plates
Description Pierce Protein A/G Coated Plate IP Kit
Content And Storage Sufficient For: Assaying 48 samples with controls or 192 total IP reactions
• Protein A/G Coated 96-well Strip Plates, 2 plates
• Uncoated 96-well Strip Plate (white), 2 plates
• PBS Pack (makes 500 mL), 1 pack
• Surfact-Amps X-100 Detergent Solution, 6 x 10 mL
• Elution Buffer (pH 2.8), 50 mL
• Neutralization Buffer, 12 mL
• Sealing Tape for 96-well Plates, 18 sheets

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microplate Mixer
Product Line Pierce
Quantity 2 plate kit
For Use With (Application) Immunoprecipitation (IP)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 192 Samples
Format Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.