Learn More
Thermo Scientific™ Pierce™ Chromatography Cartridge Accessory Pack
Catalog No. PI89971
Change view
Click to view available options
Quantity:
1 pack
Catalog No. Quantity
PI89971 1 pack
1 options
Catalog No. PI89971 Supplier Thermo Scientific™ Supplier No. 89971
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessory pack of caps and adaptors to fit 1mL and 5mL cartridges of desalting or affinity resins for manual or automated liquid chromatography equipment.
Thermo Scientific™ Pierce Chromatography Cartridges are convenient, reliable, ready-to-use, pre-packed 1mL and 5mL columns of sample-prep and affinity-purification resins for manual or automated liquid chromatography.
The cartridge-fittings are compatible with the popular automated liquid-chromatography systems or for manual syringe processing. The cartridges attach directly to AKTA™ or FPLC Systems without additional connectors. Cartridges can be used individually or connected in a series to obtain even higher column capacity. Each product supplied in the Pierce Chromatography Cartridge format includes an accessory pack that readily adapts cartridges for use with Luer-Lok™ Syringe Fittings or tubing. The cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Highlights:
  • Two sizes– 1mL and 5mL, convenient for typical research scales
  • Compatible– fittings allow connection with popular LC systems or a standard syringe
  • Versatile– use singly or connected in series to service different capacity requirements
  • Validated– available cartridges have been tested to ensure performance in the format
  • Reusable– accessory pack includes caps for convenient storage between uses
  • Economical– comparable performance at lower cost than other commercially available cartridges
Specifications
Product Type Chromatography Cartridge Accessory
Product Line Pierce
Content And Storage Sufficient For: Attaching and replacing fittings for products supplied in Pierce Chromatography Cartridges
• Bottom plug, 1
• Cap, 1
• Luer-Lok Adapter, 1
• Tube Fitting, 1
Quantity 1 pack
For Research Use Only. Not for use in diagnostic procedures.