missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CHAPS Detergent (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate)

Catalog No. pi28300
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28300 5 g
PI28299 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI28300 Supplier Thermo Scientific™ Supplier No. LSG28300
Only null left

Non-denaturing Zwitterionic detergents that disrupt nonspecific protein interactions with less protein aggregation than nonionic detergents.

Thermo Scientific™ CHAPS is a zwitterionic detergent that is especially well suited for protecting the native state of proteins.

CHAPS is a sulfobetaine derivatives of cholic acid. This zwitterionic detergent is useful for membrane protein solubilization when it is important to maintain protein activity. CHAPS detergent is soluble over a wide range of pH (2 to 12) and are easily removed from solution by dialysis because of their high critical micelle concentration (CMC).

Highlights:

  • Versatile zwitterionic detergents
  • Provide mild but effective lysis of cultured mammalian cells
  • Nondenaturing and generally do not inactivate protein functions
  • Can be removed from solution by dialysis
  • Commonly used for isoelectric focusing (IEF) and 2D electrophoresis

Properties of CHAPS detergent:

  • Chemical Name: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
  • Molecular Weight: 614.88
  • Detergent Class: Zwitterionic
  • Aggregation Number: 10
  • Micelle Molecular Weight: 6149
  • Critical Micelle Concentration (CMC): 8 to 10mM (0.4920 to 0.6150%, w/v)
  • Cloud Point: ≥ 100°C
  • Dialyzable: Yes

 

Specifications

Product Type Detergent
Content And Storage Store at Room Temperature
Form Powder
Reagent Type Detergent (Pure)
Quantity 5 g

For Research Use Only. Not for use in diagnostic procedures.