missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Centrifuge Columns

Catalog No. PI89868
Change view
Click to view available options
Capacity (Metric):
800 μL
2 mL
5 mL
10 mL
Quantity:
25 Columns
4 x 50 Columns
50 Columns
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
PI89868 800 μL 50 Columns
PI89869 800 μL 4 x 50 Columns
PI89896 2 mL 25 Columns
PI89897 5 mL 25 Columns
PI89898 10 mL 25 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI89868 Supplier Thermo Scientific™ Supplier No. 89868

Pack these empty 0.5 to 10 mL spin columns with agarose beads to perform small-scale affinity purification methods in 15- or 50-mL conical microcentrifuge tubes.

Pierce Centrifuge Columns

Thermo Scientific Pierce centrifuge columns are convenient tools for handling 40 μL to 20 mL of an affinity support for purification. The columns are made from low-protein-binding polypropylene and contain a polyethylene filter (~30-μm pore size) for compatibility with protein purification.

Simply add the affinity resin to the polypropylene column, remove the twist-off bottom, and allow the resin to pack itself. Then add your sample, allow it to bind to the support, and use a centrifuge to efficiently wash away any contaminants and elute your purified protein.

Pierce centrifuge columns can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and binding steps with dilute samples when slow processing time is desired, column extenders can be used to increase the column reservoir capacity. Use any combination of processing techniques to get the best possible chromatography results.

0.8 mL columns

  • Maximum volume: 800 μL
  • Recommended resin volume: 40–400 μL
  • Receiver tube: fits standard microcentrifuge tubes
  • Cap: O-ring screw-top
  • Twist-off bottom

2 mL columns

  • Maximum volume: 5 mL (2 mL resin bed, 3 mL sample)
  • Recommended resin volume: 400 μL–2 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

5 mL columns

  • Maximum volume: 8 mL (5 mL resin bed, 3 mL sample)
  • Recommended resin volume: 1–5 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

10 mL columns

  • Maximum volume: 22 mL (10 mL resin bed, 12 mL sample)
  • Recommended resin volume: 2–10 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

20 mL columns

  • Maximum volume: 34 mL (20 mL resin bed, 14 mL sample)
  • Recommended resin volume: 4–20 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

Applications

  • Affinity purification, affinity chromatography
  • Immuno-depletion
  • Ion exchange resins
  • Gel filtration, desalting, buffer exchange

Specifications

Bed Volume 400 μL
Capacity (Metric) 800 μL
Content And Storage Store at room temperature.
Product Line Pierce
Quantity 50 Columns
Type Spin Column
Sample Volume 800 μL
Format Microcentrifuge, Spin Column
Column Type Empty Column, Polyethylene Filter

For Research Use Only. Not for use in diagnostic procedures.