missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cell Surface Protein Isolation Kit

Catalog No. PI89881
Change view
Click to view available options
Quantity:
8 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89881 8 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89881 Supplier Thermo Scientific™ Supplier No. LSG89881
Only null left

Selectively biotinylate and purify mammalian cell surface proteins and exclude intracellular proteins with this complete set of reagents.

Thermo Scientific™ Cell Surface Protein Isolation Kit is a complete set of reagents for selective biotinylation and subsequent purification of mammalian cell surface proteins to the exclusion of intracellular proteins.

The kit efficiently labels proteins that have accessible lysine residues and sufficient extracellular exposure.The isolation procedure uses a cell-impermeable, cleavable biotinylation reagent (Sulfo-NHS-SS-Biotin) to label exposed primary amines of proteins on the surface of intact adherent or suspension cells. Treated cells are then harvested and lysed, and the labeled surface proteins are affinity-purified using Thermo Scientific NeutrAvidin Agarose Resin. The isolated cell surface proteins contain a small, nonreactive tag of the originally labeled primary amines but are no longer biotinylated (biotin remains bound to the resin). The kit contains sufficient reagents for eight experiments, each involving four T75 flasks of confluent cells.

Highlights:

  • Isolates cell surface proteins – reduces complexity of total cellular protein

  • Efficiently recovers labeled proteins – cleavable biotin allows for nearly 100% recovery of isolated cell surface proteins

  • Convenience – all reagents are provided in one kit, along with complete instructions for labeling, cell lysis and purification of cell surface membrane proteins

  • Western blotting applications – proteins recovered in SDS-PAGE buffer are loaded directly onto polyacrylamide gels

  • Robust system – protocol designed for diverse cell lines, including NIH 3T3, HeLa, C6 and A431

Includes:

Biotinylation reagent, cell lysis reagent, agarose affinity resin, various buffers and microcentrifuge spin columns

Specifications

Product Type Protein Isolation Kit
Content And Storage Upon receipt store kit at 4°C. Product shipped at ambient temperature.
Form Liquid, Powder
Product Line Pierce
Buffer Lysis Buffer
Reagent Type Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation
Sufficient For 8 Reactions
Quantity 8 reactions kit

For Research Use Only. Not for use in diagnostic procedures.