missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)

Catalog No. p-4530979
Change view
Click to view available options
Quantity:
100 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA35396 10 x 1 mg
PI21581 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA35396 Supplier Thermo Scientific™ Supplier No. A35396

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink amines to amines with these long (21.7A and 35.8A) crosslinkers composed of NHS-ester groups and 5-unit or 9-unit PEG spacer arms that enhance protein solubility.

Thermo Scientific™ Pierce BS(PEG)n Crosslinkers include BS(PEG)5 and BS(PEG)9, which are bis-succinimide ester-activated PEG compounds for crosslinking between primary amines in proteins and other molecules.

BS(PEG)5 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG5 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Features of BS(PEG)5:

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • NHS esters react with—NH2 groups at pH 7-9, forming stable amide bonds
  • Water-soluble; compare to non-pegylated BS3 (also soluble) (Cat. No. 21580) and DSS (water-insoluble) (Cat. No. 21655)
  • Membrane-impermeable, allowing for cell surface labeling
  • Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
  • Polyethylene glycol spacer arms help maintain conjugate solubility
  • Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Ideal for small molecule or peptide conjugations

Alternative Names for BS(PEG)n:

  • Bis-NHS-dPEG
  • NHS-PEG-NHS
  • BS(PEO)5
  • Bis(NHS)PEO5
  • BS-PEO9
  • dPEG NHS ester
  • Bis-succinimide ester PEG 400

Product References

  • Crosslinker Application Guide -- search for recent literature references for this product
  • Bioconjugate Techniques, Academic Press (Cat. No. 20036)

Related Products

  • BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate) (Cat. No. 21582)
  • BS3 (bis(sulfosuccinimidyl)suberate) (Cat. No. 21580)

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description BS(PEG)5
Molecular Weight (g/mol) 532.5
PEGylated Yes
Spacer Arm Length 21.7 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Wet Ice
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 10 x 1 mg
Solubility Water
Format Standard, Single-use
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)?

BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.