missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BMPH (N-β-maleimidopropionic acid hydrazide)

Catalog No. PI22297
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22297 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22297 Supplier Thermo Scientific™ Supplier No. LSG22297

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this short (8.1A) crosslinker containing maleimide and hydrazide groups.

Thermo Scientific™ Pierce BMPH is a short, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Highlights:

  • Water-soluble
  • Reactive groups: maleimide and hydrazide
  • Reactive toward: sulfhydryl groups and carbonyl aldehyde groups (or carboxyl groups when used with EDC)

Specifications

Chemical Reactivity Carbohydrate-Sulfhydryl
Cleavable No
Description BMPH
Molecular Weight (g/mol) 297.19
PEGylated No
Spacer Arm Length 8.1 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Hydrazide, Maleimide
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.