missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BMH (bismaleimidohexane)

Catalog No. PI22330
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22330 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22330 Supplier Thermo Scientific™ Supplier No. LSG22330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this long crosslinker composed of maleimide groups and 11-atom (13.0A) spacer arm.

Thermo Scientific™ Pierce BMH is a long-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).

Highlights:

  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups

Specifications

Chemical Reactivity Sulfhydryl-Sulfhydryl
Cleavable No
Description BMH
Molecular Weight (g/mol) 276.29
PEGylated No
Spacer Arm Length 13.0 Å
Content And Storage Upon receipt store at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for BMH (bismaleimidohexane)?

BMH (bismaleimidohexane) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.