missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BM(PEG)2 (1,8-bismaleimido-diethyleneglycol)

Catalog No. pi22336
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22336 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22336 Supplier Thermo Scientific™ Supplier No. LSG22336

Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, pegylated, water-soluble crosslinker composed of maleimide groups and 2-unit (14.7A) PEG spacer arm.

Thermo Scientific™ Pierce BM(PEG)2 is a pegylated crosslinker having the form maleimide-PEG-maleimide for conjugation between sulfhydryl groups (e.g., cysteines) via a two-unit polyethylene glycol (PEG) spacer arm (14.7 Ås).

Highlights:

  • Long sulfhydryl-reactive, homobifunctional crosslinker
  • Water-soluble
  • Polyethylene glycol (PEG) spacer provides increased water solubility, reducing the potential for crosslinker-caused precipitation
  • Ideal for small molecule or peptide conjugations
  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups

Specifications

Chemical Reactivity Sulfhydryl-Sulfhydryl
Cleavable No
Description BM(PEG)2
Molecular Weight (g/mol) 308.29
PEGylated Yes
Spacer Arm Length 14.7 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for BM(PEG)2 (1,8-bismaleimido-diethyleneglycol)?

BM(PEG)2 (1,8-bismaleimido-diethyleneglycol) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.