missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IPTG

Catalog No. PI34060
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34060 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34060 Supplier Thermo Scientific™ Supplier No. 34060

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect beta-galactosidase expression from vectors, plasmids, or DNA fragments with this chromogenic substrate.

Thermo Scientific™ Pierce beta-Galactosidase Precipitating Substrates is a chromogenic substrate for beta-galactosidase that is used to detect expression of vectors, plasmids, or DNA fragments that contain the reporter gene.

X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) is a chromogenic substrate forβ-Galactosidase. It yields a blue precipitate, is soluble in dimethylformamide, and can be used to detect vectors, plasmids or DNA fragments that contain the β-Galactosidase gene.

Highlights:

  • See item-specific features

X-Gal (5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside)

  • Chromogenic substrate for β-Galactosidase
  • Yields a blue precipitate
  • Soluble in DMF
  • Can be used to detect vectors, plasmids, or DNA fragments that contain the β-Galactosidase gene

IPTG (Isopropyl-β-D-thiogalactopyranoside)

  • Maximizes the expression of genes in expression vectors
  • When screening a lambda bacteriophage library with an antibody, the probability of success can be increased by preventing the expression of the fusion protein until the plaques are well established
  • By placing a nitrocellulose filter containing the inducer IPTG onto the plate and continuing growth at 37°C, the expression of the potential β-galactosidase-cDNA fusion protein can be induced after three to four hours of plaque growth; the nitrocellulose filter is then screened with antibodies
  • IPTG is also used in combination with X-Gal to detect active β-galactosidase
  • Practical in applications using Bacteriophage M13 and Igt11 vectors

Specifications

Content And Storage Store at or below -20°C.
Form Powder
Quantity 1 g
Product Type IPTG (Isopropyl β-D-thiogalactopyranoside)

For Research Use Only. Not for use in diagnostic procedures.