missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ BCA Protein Assay Reagent A

Estimate visually, or measure with a standard spectrophotometer or plate reader at 562 nm.

Supplier:  Thermo Scientific™ 23221

 View more versions of this product

Catalog No. PI23221

Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce BCA Protein Assay Kit remains one of the most popular protein quantitation methods worldwide.

Additional Information and Online Ordering

  • A component of the Pierce BCA Protein Assay Kit
  • Kit is a two-component, high-precision, detergent-compatible assay that is used for total protein concentration determination compared to a protein standard
  • Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research
  • The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications
  • Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy
  • Estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
  • Exhibits less protein-to-protein variation than dye-binding methods (Bradford)
  • Unaffected by typical concentrations of most ionic and non-ionic detergents
  • Assay has a 30 min. incubation; much easier and four times faster than classical Lowry methods
  • Linear working range for BSA of 20 to 2000 μg/mL
  • Detect down to 5 μg/mL with the enhanced protocol
  • Store at room temperature
Specifications

Specifications

Store at room temperature.
BCA Assay
Pierce
Not Target-Specific
Spectrophotometer, Microplate Reader
Pierce BCA Protein Assay Reagent A
Colorimetric
Protein Quantitation Assay
Solution-based Detection, Absorbance
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.