missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ BCA Protein Assay Reagent A

Catalog No. p-9023018
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3.75 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23222 3.75 L
PI23221 250 mL
PI23223 1 L
PI23228 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI23222 Supplier Thermo Scientific™ Supplier No. 23222
Only null left

Estimate visually, or measure with a standard spectrophotometer or plate reader at 562 nm.

Thermo Scientific™ Pierce BCA Protein Assay Kit remains one of the most popular protein quantitation methods worldwide.

Additional Information and Online Ordering

  • A component of the Pierce BCA Protein Assay Kit
  • Kit is a two-component, high-precision, detergent-compatible assay that is used for total protein concentration determination compared to a protein standard
  • Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research
  • The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications
  • Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy
  • Estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
  • Exhibits less protein-to-protein variation than dye-binding methods (Bradford)
  • Unaffected by typical concentrations of most ionic and non-ionic detergents
  • Assay has a 30 min. incubation; much easier and four times faster than classical Lowry methods
  • Linear working range for BSA of 20 to 2000 μg/mL
  • Detect down to 5 μg/mL with the enhanced protocol
  • Store at room temperature

Specifications

Content And Storage Store at room temperature.
Description Pierce BCA Protein Assay Reagent A
Assay BCA Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 3.75 L

For Research Use Only. Not for use in diagnostic procedures.