Learn More
Thermo Scientific™ Pierce™ Avidin Agarose
Description
Thermo Scientific™ Pierce Avidin Agarose is high-quality beaded agarose resin for biotin affinity procedures.
Additional Information and Online Ordering
Avidin is a tetrameric glycoprotein rich in tryptophan that is present in chicken egg white and the tissues of birds, and avidin is highly resistant to denaturation by acids or proteolytic enzymes. Avidin Agarose Resin consists of purified avidin that has been immobilized to crosslinked 6% beaded agarose. The resulting affinity support is leak-resistant, stable at pH from 2 to 11, and ideal for gravity-flow and low-speed centrifugation applications.
Highlights:
- Immobilized ligand is purified avidin from chicken egg whites
- Support is crosslinked 6% beaded agarose resin (agarose beads)
- Supplied as a stable aqueous slurry or pre-packed columns
- Binds free biotin and biotinylated molecules with high affinity
Recommended for:
Binding biotinylated anti-transferrin from serum; Binding biotinyl peptides and elution with an SDS/urea solution; Hybridization of biotinylated RNA to its complementary DNA and binding to immobilized avidin, with subsequent elution of the single-stranded DNA; Purification of double-stranded DNA
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Pierce |
| Quantity | 5 x 5 mL |
| Target | Biotin-Tagged Molecule |
| Type | Agarose |
| Stationary Phase | Avidin |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
| Binding Capacity | 20 μg Biotin Binding/mL Resin |
For Research Use Only. Not for use in diagnostic procedures.