missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Aquaporin Z Membrane Protein Standard

Catalog No. FERA66693
Change view
Click to view available options
Quantity:
20 μg
5 x 20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA66693 20 μg
FERA66695 5 x 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERA66693 Supplier Thermo Scientific™ Supplier No. A66693

The Thermo Scientific Pierce Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.

The Thermo Scientific Pierce Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.

Features of Pierce Aquaporin Z Membrane Protein Standard include:

  • Positive control sample—aquaporin Z protein production optimized and validated for use in native mass spectrometry
  • Rigorously tested—high quality, consistent protein standard documented via lot-specific certificate of analysis
  • Convenient—ready-to-use format with each single-use vial sufficient for 5 injections

Achieve confidence in your methods without wasting precious samples

Method development for membrane protein analysis provides unique challenges. Pierce Aquaporin Z Membrane Protein Standard empowers success by letting you test parameters and troubleshoot issues without wasting irreplaceable samples. This high purity standard contains sufficient material for multiple qualitative experiments by direct infusion or LC/MS. You can use it to verify system performance, tune parameters, and develop native LC/MS methods without worrying about depleting your real samples.

Specifications

Description Tetramer
Formulation Liquid in 0.05% LDAO, 200 mM Ammonium Acetate; pH 7.4
For Use With (Equipment) Q-Exactive™ UHMR, HMRn option on Tribrid Series MS systems
Material Aquaporin Z
Product Type Protein Standard
Quantity 20 μg
Color Colorless
Grade Mass Spec Grade
Molecular Weight (g/mol) 98,120 Da
Form Liquid

For Research Use Only.