Learn More
Thermo Scientific™ Pierce™ Alkylating Reagents
Description
Thermo Scientific Pierce Iodoacetamide or Chloroacetamide are provided in a no-weigh format, where small amounts of reagent are pre-weighed and packaged for single use. This single-use format prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. The format enables use of a fresh vial of reagent each time, and eliminates the hassle of weighing out small amounts of reagent.
Specifications
Specifications
| Product Type | Alkylating Reagent |
| Quantity | 10 x 2 mg |
| Includes | Chloroacetamide |
| Product Line | Pierce |
Frequently Asked Questions (FAQs)
The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html