missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ChIP-Grade Protein A/G Plus Agarose

Catalog No. PI26159
Change view
Click to view available options
Quantity:
0.65 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26159 0.65 mL
PI26161 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI26159 Supplier Thermo Scientific™ Supplier No. LSG26159
Only null left

Chromatin immunoprecipitation (chromatin IP, ChIP) assay kit containing all reagents, agarose affinity beads, and a simple, fast, reliable protocol.

Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.

This chromatin IP kit contains sufficient reagents to perform 30 ChIP assays with appropriate controls using an optimized protocol and a convenient spin-column format. The kit includes a complete Chromatin Prep Module (also available separately) to efficiently extract and solubilize crosslinked DNA-protein interaction complexes from cells, plus an immunoprecipitation module that contains high-capacity, ChIP-grade Protein A/G agarose.

Highlights:

  • Simple and streamlined ChIP assay protocol (< 8 hours)
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-capacity Protein A/G agarose resin
  • Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
  • Fast and reproducible spin-column format; columns and collection tubes included
  • High-recovery DNA purification
  • Choose a complete ChIP assay kit or the separate chromatin preparation module

Includes:

Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls

Specifications

Binding Capacity >50 mg Human IgG/mL resin binding capacity
Capacity (Metric) ≥50 mg Human IgG per mL Resin Binding
Content And Storage Upon receipt store at 4°C
Form Liquid Suspension
Product Line Pierce
Quantity 0.65 mL
Target IgG Antibody
Type Agarose
Stationary Phase Protein A/G
Format Bottle
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.