missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agarose II (Low Melt)

Catalog No. PI17856
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17856 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17856 Supplier Thermo Scientific™ Supplier No. LSG17856

Ensure uniform lot-to-lot consistency in preparing electrophoresis gels using highly purified and blended regular- or low-melting agarose formulations.

Thermo Scientific™ Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lot consistency for preparation of electrophoresis gels.

Highlights:

  • Agarose I (Molecular Biology Grade)
  • A regular melting-temperature agarose for electrophoresis requiring an exceptional level of purity with unequaled performance
  • Agarose II (Low Melt)
  • A low temperature agarose for special applications requiring a lower melting temperature
  • Optimal for “in-gel” restriction digestion/ligation and DNA recovery
  • At 1%, forms a very manageable clear gel that provides excellent resolution and melting characteristics
  • Melts at approximately 65°C and remains fluid at 37°C for at least 24 hours, providing an excellent matrix for enzymatic manipulation of samples

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Gel Electrophoresis
Melting Point Low Melting Point
Product Type Agarose II (Low Melt)
Quantity 100 g
Separation Range 100 bp to >30 kb