missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 8M Guanidine-HCl Solution

Catalog No. PI24115
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24115 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI24115 Supplier Thermo Scientific™ Supplier No. LSG24115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use, 8M solution of guanidinium hydrochloride (GdnCl, GuHCl), CAS 50-01-1; dilute and pH-adjust to any concentration for protein biology methods.

Thermo Scientific™ Pierce 8M Guanidine-HCl Solution is an accurately formulated, ready-to-use solution of guanidine hydrochloride, that can be easily diluted and pH-adjusted to any concentration below 8M.

Highlights:

  • Any concentration below 8 M can be quickly and easily prepared
  • No more wasting valuable research time handling difficult-to-weigh crystals for slow-to-prepare guanidine·HCl solutions
  • Free of UV-absorbing materials at 225-300nm
  • Sharp UV cut-off spectrum with OD260 < 0.03
  • Typical metals: Cu < 0.02ppm, Zn < 0.15ppm, Fe < 0.25ppm, Pb < 0.005ppm
  • Particulate-free, crystal-clear solution
  • Aldehyde-free
  • Excellent stability

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Form Liquid
Reagent Type Chaotrope, Denaturant
Quantity 200 mL
Product Type Guanidine Hydrochloride
What is the expiration date for the 8M Guanidine-HCl Solution (Cat. No. 24115) and is its stability affected by opening the bottle?

There is no expiration date for the 8M Guanidine-HCl Solution (Cat. No. 24115). Stability is not affected by opening the bottle.

For Research Use Only. Not for use in diagnostic procedures.