missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Saturated, Reagents

Catalog No. p-200021518
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334276 1 L
01334275 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334276 Supplier Labchem Inc Supplier No. CP1224001A
Only null left

Picric Acid, Saturated, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2,4,6-Trinitrophenol
IUPAC Name water
SMILES O

Specifications

pH 1.2 to 2
Boiling Point 100°C
Color Yellow
Grade Reagent
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0074
Chemical Name or Material Picric Acid, Saturated