missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Saturated, Approximately 1.2% (w/v), Ricca Chemical

Catalog No. p-3252079
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
500 mL HDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01213112 100 mL 500 mL HDPE Bottle
586016 500 mL Natural Poly Bottle
586032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01213112 Supplier Ricca Chemical Company Supplier No. R5860000100A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol

Specifications

Melting Point 0.0°C
Boiling Point 100°C
Color Yellow
CAS Max % 1.18%, 98.82%
CAS Min % 1.18%, 98.82%
Appearance Yellow Liquid
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Grade Laboratory
Quantity 100 mL
Packaging 500 mL HDPE Bottle
Physical Form Liquid
Chemical Name or Material Picric Acid
Show More Show Less
ShelfLife 24 Months